trans-N-Fmoc-4-tert-butoxy-L-proline 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-4-trans-(tert-butoxy)-L-proline, Fmoc-Hyp(tBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-trans-(tert-butoxy)-L-proline, (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxy)pyrrolidine-2-carboxylic acid
trans-N-Fmoc-4-tert-butoxy-L-proline, 98%
Purity: 98%
CAS No.: 122996-47-8
Molecular Formula: C24H27NO5
Formula Weight: 409.47
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 6161467
Optical Rotation: -25°(c=1 in methanol)
MDL No.: MFCD00151930
Formula:
C₂₄H₂₇NO₅ MW: 409.482 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151930 CAS Number: 122996-47-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...