2-Methylquinoline-6-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63790-03
H63790-06
H63790-14
AAH63790-03EA
29.07
CAD
AAH63790-03
AAH63790-06
AAH63790-14
2-Methylquinoline-6-carboxylic acid 97%
2-Methylquinoline-6-carboxylic acid
2-Methylquinoline-6-carboxylic acid, 97%
Purity: 97%
CAS No.: 635-80-3
Molecular Formula: C11H9NO2
Formula Weight: 187.19
Physical Form: Solid
MDL No.: MFCD00014566
Formula:
C11H9NO2 MW: 187.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014566 CAS Number: 635-80-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...