3-Iodo-L-tyrosine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
H63789-03
H63789-06
AAH63789-03EA
72.84
CAD
AAH63789-03
AAH63789-06
3-Iodo-L-tyrosine 98%
3-Iodo-L-tyrosine
3-Iodo-L-tyrosine, 98%
Purity: 98%
CAS No.: 70-78-0
Molecular Formula: C9H10INO3
Formula Weight: 307.09
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 210° dec
Beilstein Reference: 2941266
MDL No.: MFCD00002608
Formula:
C₉H₁₀INO₃ MW: 307.1 g/mol Melting Pt: 210 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002608 CAS Number: 70-78-0 EINECS: 200-744-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...