Boc-Phe-Phe-OH
Supplier: Thermo Scientific Chemicals
H63788-03
H63788-06
AAH63788-03EA
85.64
CAD
AAH63788-03
AAH63788-06
Boc-Phe-Phe-OH
Proteins and Peptides
N-Boc-L-phenylalanyl-L-phenylalanine, 95%
Purity: 95%
CAS No.: 13122-90-2
Molecular Formula: C23H28N2O5
Formula Weight: 412.48
Physical Form: Solid
MDL No.: MFCD00190824
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...