L-Valinamide hydrochloride 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Val-NH2.HCl, H-Val-NH₂·HCl
H63775-06
H63775-14
H63775-22
AAH63775-06EA
65.73
CAD
AAH63775-06
AAH63775-14
AAH63775-22
L-Valinamide hydrochloride 95%
L-Valinamide hydrochloride
L-Valinamide hydrochloride, 95%
Purity: 95%
CAS No.: 3014-80-0
Molecular Formula: C5H12N2O.HCl
Formula Weight: 152.62
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 266-270°
Beilstein Reference: 4198583
Optical Rotation: +26°(c=1 in water)
MDL No.: MFCD00039085
Formula:
C₅H₁₂N₂O MW: 152.62 g/mol Melting Pt: 266…270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039085 CAS Number: 3014-80-0 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...