Fmoc-aspartic acid-4-methyl ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Fmoc-L-aspartic acid 4-methyl ester, Fmoc-Asp(OMe)-OH

H63774-03 H63774-06
AAH63774-03EA 60.36 CAD
AAH63774-03 AAH63774-06 AAH63774-14
Fmoc-aspartic acid-4-methyl ester 97%
Fmoc-aspartic acid-4-methyl ester

N-Fmoc-L-aspartic acid 4-methyl ester, 97%


Purity: 97% CAS No.: 145038-53-5 Molecular Formula: C20H19NO6 Formula Weight: 369.37 Physical Form: Solid MDL No.: MFCD08458573

Formula: C₂₀H₁₉NO₆
MW: 369.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458573
CAS Number: 145038-53-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR