Fmoc-Lys(Alloc)-OH 95%
Supplier: Thermo Scientific Chemicals
H63768-03
H63768-06
AAH63768-03EA
58.62
CAD
AAH63768-03
AAH63768-06
Fmoc-Lys(Alloc)-OH 95%
Fmoc-Lys(Alloc)-OH
N^e-Allyloxycarbonyl-N^a-Fmoc-L-lysine, 95%
Purity: 95%
CAS No.: 146982-27-6
Molecular Formula: C25H28N2O6
Formula Weight: 452.50
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 5893081
Optical Rotation: -12°(c=1 in DMF)
MDL No.: MFCD00190872
Formula:
C₂₅H₂₈N₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00190872 CAS Number: 146982-27-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...