2,3-Dichloroisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dichloropyridine-4-carboxylic acid

H63733-03 H63733-06
AAH63733-03EA 136.2 CAD
AAH63733-03 AAH63733-06
2,3-Dichloroisonicotinic acid 97%
2,3-Dichloroisonicotinic acid

2,3-Dichloropyridine-4-carboxylic acid, 97%


Purity: 97% CAS No.: 184416-84-0 Molecular Formula: C6H3Cl2NO2 Formula Weight: 192.00 Physical Form: Solid Melting Point: 203-207° MDL No.: MFCD01861977

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 203…207 °C
Storage Temperature: Ambient
MDL Number: MFCD01861977
CAS Number: 184416-84-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR