H-Lys(boc)-OMe·HCl 98%
Supplier: Thermo Scientific Chemicals
H63727-03
H63727-06
AAH63727-03EA
79.79
CAD
AAH63727-03
AAH63727-06
H-Lys(boc)-OMe·HCl 98%
H-Lys(boc)-OMe·HCl
N^e-Boc-L-lysine methyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 2389-48-2
Molecular Formula: C12H24N2O4.HCl
Formula Weight: 296.79
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 5173577
MDL No.: MFCD00076959
Formula:
C₁₂H₂₅ClN₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00076959 CAS Number: 2389-48-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...