(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(((benzyloxy)methyl)amino)butanoic acid, Fmoc-Dab(Z)-OH

H63726-03 H63726-06
AAH63726-03EA 180.12 CAD
AAH63726-03 AAH63726-06 AAH63726-MD
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid 95%
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.


Purity: 95% CAS No.: 252049-08-4 Molecular Formula: C27H26N2O6 Formula Weight: 474.51 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD02259487

Formula: C₂₇H₂₈N₂O₅
MW: 460.53 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02259487
CAS Number: 252049-08-4

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR