Methyl-3-amino-5-methylthiophene-2-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-amino-5-methylthiophene-2-carboxylate

H63719-03 H63719-06
AAH63719-03EA 257.54 CAD
AAH63719-03 AAH63719-06 AAH63719-MD
Methyl-3-amino-5-methylthiophene-2-carboxylate 97%
Methyl-3-amino-5-methylthiophene-2-carboxylate

Methyl 3-amino-5-methylthiophene-2-carboxylate, 97%


Purity: 97% CAS No.: 76575-71-8 Molecular Formula: C7H9NO2S Formula Weight: 171.22 Physical Form: Solid MDL No.: MFCD00130095

Formula: C₇H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00130095
CAS Number: 76575-71-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR