2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Trichloroacetyl)pyrrole, 2-(trichloroacetyl)-1h-pyrrole
H63718-06
H63718-14
AAH63718-06EA
62.73
CAD
AAH63718-06
AAH63718-14
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone 99+%
2,2,2-Trichloro-1-(1H-pyrrol-2-yl)ethanone
2-(Trichloroacetyl)pyrrole
Purity: 99+%
CAS No.: 35302-72-8
Molecular Formula: C6H4Cl3NO
Formula Weight: 212.46
Physical Form: Powder
Appearance: Pale grey
Melting Point: 72-74°
Beilstein Reference: 135479
MDL No.: MFCD00128757
Formula:
C₆H₄Cl₃NO MW: 212.46 g/mol Melting Pt: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128757 CAS Number: 35302-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...