5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Formyl-2,4-dimethyl-3-pyrrolecarboxylic acid, 5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid
H63711-03
H63711-06
H63711-MD
AAH63711-03EA
136.51
CAD
AAH63711-03
AAH63711-06
AAH63711-MD
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 96%
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
Purity: 96%
CAS No.: 253870-02-9
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Physical Form: Solid
MDL No.: MFCD06202342
Formula:
C8H9NO3 Storage Temperature: Ambient |
MDL Number:
MFCD06202342 CAS Number: 253870-02-9 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...