H-Glu(OtBu)-OtBu·HCl 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Glu(OtBu)-OtBu·HCl, L-Glutamic acid di-tert-butyl ester hydrochloride, H-Glu(OtBu)-OtBu.HCl
H63708-03
H63708-06
H63708-14
AAH63708-03EA
67.78
CAD
AAH63708-03
AAH63708-06
AAH63708-14
H-Glu(OtBu)-OtBu·HCl 98%
H-Glu(OtBu)-OtBu·HCl
L-Glutamic acid di-tert-butyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 32677-01-3
Molecular Formula: C13H25NO4.HCl
Formula Weight: 295.8
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 3573738
MDL No.: MFCD00058003
Formula:
C₁₃H₂₆ClNO₄ MW: 295.804 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00058003 CAS Number: 32677-01-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...