1-Phenyl-1H-pyrazole-4-carbaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63695-03
H63695-MD
AAH63695-03EA
230.68
CAD
AAH63695-03
AAH63695-MD
1-Phenyl-1H-pyrazole-4-carbaldehyde 98%
1-Phenyl-1H-pyrazole-4-carbaldehyde
1-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%
Purity: 98%
CAS No.: 54605-72-0
Molecular Formula: C10H8N2O
Formula Weight: 172.18
Physical Form: Solid
Sensitivity: Moisture and Light Sensitive
Appearance: Pale yellow
Melting Point: 84-88°
MDL No.: MFCD02179568
Formula:
C₁₀H₈N₂O Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179568 CAS Number: 54605-72-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...