Fmoc-3-(1-naphthyl)-D-alanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-D-1-Nal-OH, N-Fmoc-3-(1-naphthyl)-D-alanine
H63688-03
H63688-06
AAH63688-03EA
84.53
CAD
AAH63688-03
AAH63688-06
AAH63688-MD
Fmoc-3-(1-naphthyl)-D-alanine 98%
Fmoc-3-(1-naphthyl)-D-alanine
N-Fmoc-3-(1-naphthyl)-D-alanine, 98%
Purity: 98%
CAS No.: 138774-93-3
Molecular Formula: C28H23NO4
Formula Weight: 437.49
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 9160026
MDL No.: MFCD00191200
Formula:
C₂₈H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00191200 CAS Number: 138774-93-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...