Boc-Abu-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-(tert-Butoxycarbonylamino)butyric acid, Boc-L-2-aminobutanoic acid, BOC-L-α-Aminobutyric acid, Boc-L-Abu-OH, (S)-2-(Boc-amino)butyric acid
H63686-03
H63686-06
H63686-14
AAH63686-03EA
68.41
CAD
AAH63686-03
AAH63686-06
AAH63686-14
Boc-Abu-OH 95%
Boc-Abu-OH
(S)-2-(Boc-amino)butyric acid, 95%
Purity: 95%
CAS No.: 34306-42-8
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Physical Form: Solid
Beilstein Reference: 6801706
Optical Rotation: -18°(c=1 in methanol)
MDL No.: MFCD00037267
Formula:
C₉H₁₇NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00037267 CAS Number: 34306-42-8 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...