(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
H-D-Ser-OMe·HCl, H-D-Ser-OMe.HCl, D-Serine methyl ester hydrochloride
H63685-06
H63685-14
H63685-22
AAH63685-06EA
44.24
CAD
AAH63685-06
AAH63685-14
AAH63685-22
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride
D-Serine methyl ester hydrochloride, 95%
Purity: 95%
CAS No.: 5874-57-7
Molecular Formula: C4H9NO3.HCl
Formula Weight: 155.58
Physical Form: Solid
Melting Point: 163-166° dec
Optical Rotation: -4°(c=4 in ethanol)
MDL No.: MFCD00066121
Formula:
C4H10ClNO3 Melting Pt: 163…166 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066121 CAS Number: 5874-57-7 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...