O-Benzyl-L-tyrosine 98%
Supplier: Thermo Scientific Chemicals
H63675-06
H63675-14
AAH63675-06EA
102.07
CAD
AAH63675-06
AAH63675-14
AAH63675-22
O-Benzyl-L-tyrosine 98%
O-Benzyl-L-tyrosine
O-Benzyl-L-tyrosine, 98%
Purity: 98%
CAS No.: 16652-64-5
Molecular Formula: C16H17NO3
Formula Weight: 271.31
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 259° dec
Beilstein Reference: 2659642
Optical Rotation: -10°(c=1 in acetic acid: water (4:1))
MDL No.: MFCD00002605
Formula:
C₁₆H₁₇NO₃ Melting Pt: 259 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002605 CAS Number: 16652-64-5 EINECS: 240-699-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...