Fmoc-N-Me-Val-OH 95%
Supplier: Thermo Scientific Chemicals
H63671-03
H63671-06
AAH63671-03EA
142.83
CAD
AAH63671-03
AAH63671-06
Fmoc-N-Me-Val-OH 95%
Fmoc-N-Me-Val-OH
N-Fmoc-N-methyl-L-valine, 95%
Purity: 95%
CAS No.: 84000-11-3
Molecular Formula: C21H23NO4
Formula Weight: 353.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 187-190°
Optical Rotation: -68°(c=1 in DMF)
MDL No.: MFCD00153395
Formula:
C₂₁H₂₃NO₄ Melting Pt: 187…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153395 CAS Number: 84000-11-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...