Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
H63666-06
H63666-14
AAH63666-06EA
176.49
CAD
AAH63666-06
AAH63666-14
Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Nomega-Nitro-L-arginine methyl ester hydrochloride
N^w-Nitro-L-arginine methyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 51298-62-5
Molecular Formula: C7H15N5O4.HCl
Formula Weight: 269.69
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 159-161°
Beilstein Reference: 3744166
Optical Rotation: +15°(c=3 in methanol)
MDL No.: MFCD00039052
Formula:
C₇H₁₅N₅O₄ Melting Pt: 159…161 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039052 CAS Number: 51298-62-5 EINECS: 257-116-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...