5-tert-Butyl-1-methyl-L-glutamate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester Hydrochloride, H-Glu(OtBu)-OMe·HCl, H-Glu(OtBu)-OMe.HCl
H63661-03
H63661-06
AAH63661-03EA
114.08
CAD
AAH63661-03
AAH63661-06
5-tert-Butyl-1-methyl-L-glutamate hydrochloride 98%
5-tert-Butyl-1-methyl-L-glutamate hydrochloride
L-Glutamic acid 5-tert-butyl 1-methyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 6234-01-1
Molecular Formula: C10H19NO4.HCl
Formula Weight: 253.72
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 3572140
MDL No.: MFCD00153438
Formula:
C₁₀H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00153438 CAS Number: 6234-01-1 EINECS: 228-348-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...