5-Bromotryptamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Aminoethyl)-5-bromoindole

H63653-03 H63653-06 H63653-MD
AAH63653-03EA 377.62 CAD
AAH63653-03 AAH63653-06 AAH63653-MD
5-Bromotryptamine 97%
5-Bromotryptamine

3-(2-Aminoethyl)-5-bromoindole, 97%


Purity: 97% CAS No.: 3610-42-2 Molecular Formula: C10H11BrN2 Formula Weight: 239.11 Physical Form: Solid MDL No.: MFCD00130169

Formula: C₁₀H₁₁BrN₂
MW: 239.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130169
CAS Number: 3610-42-2
EINECS: 222-779-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR