N-(tert-Butoxycarbonyl)-D-threonine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-D-threonine, Boc-D-Thr-OH, (2R,3S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid
H63647-06
H63647-14
AAH63647-06EA
151.84
CAD
AAH63647-06
AAH63647-14
N-(tert-Butoxycarbonyl)-D-threonine 95%
N-(tert-Butoxycarbonyl)-D-threonine
N-Boc-D-threonine, 95%
Purity: 95%
CAS No.: 55674-67-4
Molecular Formula: C9H17NO5
Formula Weight: 219.23
Physical Form: Solid
Beilstein Reference: 2051734
MDL No.: MFCD00037807
Formula:
C₉H₁₇NO₅ MW: 219.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037807 CAS Number: 55674-67-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...