D-Tyrosine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-D-Tyr-OMe.HCl, H-D-Tyr-OMe·HCl

H63630-06 H63630-14
AAH63630-06EA 80.26 CAD
AAH63630-06 AAH63630-14
D-Tyrosine methyl ester hydrochloride 98%
D-Tyrosine methyl ester hydrochloride

D-Tyrosine methyl ester hydrochloride, 98%


Purity: 98% CAS No.: 3728-20-9 Molecular Formula: C10H13NO3.HCl Formula Weight: 231.68 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00066124

Formula: C₁₀H₁₄ClNO₃
MW: 231.676 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066124
CAS Number: 3728-20-9
EINECS: 223-084-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR