Fmoc-Lys(Mtt)-OH 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine, Nα-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
H63629-03
H63629-06
AAH63629-03EA
115.18
CAD
AAH63629-03
AAH63629-06
Fmoc-Lys(Mtt)-OH 95%
Fmoc-Lys(Mtt)-OH
N^a-Fmoc-N^e-(4-methyltrityl)-L-lysine, 95%
Purity: 95%
CAS No.: 167393-62-6
Molecular Formula: C41H40N2O4
Formula Weight: 624.77
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00237166
Formula:
C₄₁H₄₀N₂O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00237166 CAS Number: 167393-62-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...