2-Thiazolecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiazole-2-carboxylic acid

H63620-03 H63620-06 H63620-MD
AAH63620-03EA 127.66 CAD
AAH63620-03 AAH63620-06 AAH63620-MD
2-Thiazolecarboxylic acid 95%
2-Thiazolecarboxylic acid

Thiazole-2-carboxylic acid, 95%


Purity: 95% CAS No.: 14190-59-1 Molecular Formula: C4H3NO2S Formula Weight: 129.14 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD02094169

Formula: C₄H₃NO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD02094169
CAS Number: 14190-59-1

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR