Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride 98%

Supplier: Thermo Scientific Chemicals

H63611-06 H63611-14
AAH63611-06EA 55.62 CAD
AAH63611-06 AAH63611-14 AAH63611-22
Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride 98%
Nalpha-Boc-L-arginine hydrochloride

N^a-Boc-L-arginine hydrochloride, 98%


Purity: 98% CAS No.: 35897-34-8 Molecular Formula: C11H22N4O4.HCl Formula Weight: 310.78 Physical Form: Powder Appearance: White Beilstein Reference: 3658939 MDL No.: MFCD00063594

Formula: C₁₁H₂₂N₄O₄·HCl
Storage Temperature: Ambient
MDL Number: MFCD00063594
CAS Number: 35897-34-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR