Fmoc-Lys(biotin)-OH 95%
Supplier: Thermo Scientific Chemicals
H63609-03
H63609-06
AAH63609-03EA
455.04
CAD
AAH63609-03
AAH63609-06
AAH63609-MD
Fmoc-Lys(biotin)-OH 95%
Fmoc-Lys(biotin)-OH
N^a-Biotinyl-N^e-Fmoc-L-lysine, 95%
Purity: 95%
CAS No.: 146987-10-2
Molecular Formula: C31H38N4O6S
Formula Weight: 594.72
Physical Form: Solid
MDL No.: MFCD00270741
Formula:
C₃₁H₃₈N₄O₆S Storage Temperature: Ambient |
MDL Number:
MFCD00270741 CAS Number: 146987-10-2 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...