6-Bromo-2,4-dichloroquinazoline 97%

Supplier: Thermo Scientific Chemicals
Warning

H63603-03 H63603-06
AAH63603-03EA 230.68 CAD
AAH63603-03 AAH63603-06 AAH63603-MD
6-Bromo-2,4-dichloroquinazoline 97%
6-Bromo-2,4-dichloroquinazoline

6-Bromo-2,4-dichloroquinazoline, 97%


Purity: 97% CAS No.: 102393-82-8 Molecular Formula: C8H3BrCl2N2 Formula Weight: 277.93 Physical Form: Solid Appearance: Off-white MDL No.: MFCD09744007

Formula: C₈H₃BrCl₂N₂
MW: 277.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09744007
CAS Number: 102393-82-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR