N-Boc-thiourea 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl carbamothioylcarbamate

H63599-03 H63599-06 H63599-MD
AAH63599-03EA 138.88 CAD
AAH63599-03 AAH63599-06 AAH63599-MD
N-Boc-thiourea 97%
N-Boc-thiourea

N-Boc-thiourea, 97%


Purity: 97% CAS No.: 268551-65-1 Molecular Formula: C6H12N2O2S Formula Weight: 176 Physical Form: Powder Appearance: Off-white Melting Point: 142° dec MDL No.: MFCD04039987

Formula: C₆H₁₂N₂O₂S
MW: 176.234 g/mol
Melting Pt: 142 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04039987
CAS Number: 268551-65-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR