Fmoc-Chg-OH 98%
Supplier: Thermo Scientific Chemicals
H63589-03
H63589-06
H63589-MD
AAH63589-03EA
128.14
CAD
AAH63589-03
AAH63589-06
AAH63589-MD
Fmoc-Chg-OH 98%
Fmoc-Chg-OH
N-Fmoc-2-cyclohexyl-L-glycine, 98%
Purity: 98%
CAS No.: 161321-36-4
Molecular Formula: C23H25NO4
Formula Weight: 379.45
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00190878
Formula:
C₂₃H₂₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00190878 CAS Number: 161321-36-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...