4-(Boc-amino)indole-3-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63587-03
H63587-MD
AAH63587-03EA
824.76
CAD
AAH63587-03
AAH63587-MD
4-(Boc-amino)indole-3-carboxaldehyde 97%
4-(Boc-amino)indole-3-carboxaldehyde
4-(Boc-amino)indole-3-carboxaldehyde, 97%
Purity: 97%
CAS No.: 885266-77-3
Molecular Formula: C14H16N2O3
Formula Weight: 260.29
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD04973990
Formula:
C₁₄H₁₆N₂O₃ MW: 260.293 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973990 CAS Number: 885266-77-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...