L-1-Naphthylalanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-1-Nal-OH, 3-(1-Naphthyl)-L-alanine
H63586-03
H63586-06
AAH63586-03EA
82
CAD
AAH63586-03
AAH63586-06
AAH63586-MD
L-1-Naphthylalanine 95%
L-1-Naphthylalanine
3-(1-Naphthyl)-L-alanine, 95%
Purity: 95%
CAS No.: 55516-54-6
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 4352833
MDL No.: MFCD00066086
Formula:
C₁₃H₁₃NO₂ MW: 215.252 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066086 CAS Number: 55516-54-6 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...