Fmoc-Asp(OAll)-OH 98%
Supplier: Thermo Scientific Chemicals
H63585-03
H63585-06
H63585-14
AAH63585-03EA
50.72
CAD
AAH63585-03
AAH63585-06
AAH63585-14
Fmoc-Asp(OAll)-OH 98%
Fmoc-Asp(OAll)-OH
N-Fmoc-L-aspartic acid 4-allyl ester, 98%
Purity: 98%
CAS No.: 146982-24-3
Molecular Formula: C22H21NO6
Formula Weight: 395.41
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 111-115°
Beilstein Reference: 6670219
Optical Rotation: -27°(c=1 in DMF)
MDL No.: MFCD00190874
Formula:
C22H21NO6 Melting Pt: 111…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190874 CAS Number: 146982-24-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...