2-Oxa-6-azaspiro[3.3]heptane oxalate 97%

Supplier: Thermo Scientific Chemicals
Warning

H63578-03 H63578-06 H63578-MD
AAH63578-03EA 334.96 CAD
AAH63578-03 AAH63578-06 AAH63578-MD
2-Oxa-6-azaspiro[3.3]heptane oxalate 97%
2-Oxa-6-azaspiro[3.3]heptane oxalate

2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%


Purity: 97% CAS No.: 1159599-99-1 Molecular Formula: C5H9NO.C2H2O4 Formula Weight: 189.17 MDL No.: MFCD11976178

Formula: C₇H₁₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD11976178
CAS Number: 1159599-99-1

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR