4-Chloro-L-phenylalanine 97%

Supplier: Thermo Scientific Chemicals

H63573-03 H63573-06 H63573-MD
AAH63573-03EA 285.98 CAD
AAH63573-03 AAH63573-06 AAH63573-MD
4-Chloro-L-phenylalanine 97%
4-Chloro-L-phenylalanine

4-Chloro-L-phenylalanine, 97%


Purity: 97% CAS No.: 14173-39-8 Molecular Formula: C9H10ClNO2 Formula Weight: 199.63 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 235-238° Beilstein Reference: 2416150 UN No.: 2811 MDL No.: MFCD00063065

Formula: C₉H₁₀ClNO₂
Melting Pt: 235…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063065
CAS Number: 14173-39-8
EINECS: 238-023-5
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR