N-Succinimidyl N-methylcarbamate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63566-03
H63566-06
H63566-14
AAH63566-03EA
70.15
CAD
AAH63566-03
AAH63566-06
AAH63566-14
N-Succinimidyl N-methylcarbamate 97%
N-Succinimidyl N-methylcarbamate
N-Succinimidyl N-methylcarbamate, 97%
Purity: 97%
CAS No.: 18342-66-0
Molecular Formula: C6H8N2O4
Formula Weight: 172.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Beilstein Reference: 1531863
MDL No.: MFCD00800292
Formula:
C₆H₈N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00800292 CAS Number: 18342-66-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...