4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
Supplier: Thermo Scientific Chemicals
H63554-06
H63554-14
AAH63554-06EA
85.16
CAD
AAH63554-06
AAH63554-14
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate
N-Boc-L-aspartic acid 4-tert-butyl ester, 98%
Purity: 98%
CAS No.: 1676-90-0
Molecular Formula: C13H23NO6
Formula Weight: 289.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 64-67°
Beilstein Reference: 2336144
Optical Rotation: +2°(c=1 in methanol)
MDL No.: MFCD00076912
Formula:
C₁₃H₂₃NO₆ Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076912 CAS Number: 1676-90-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...