6-Bromoquinazoline-2,4(1H,3H)-dione 97%

Supplier: Thermo Scientific Chemicals
Warning

H63547-03 H63547-06
AAH63547-03EA 104.44 CAD
AAH63547-03 AAH63547-06 AAH63547-MD
6-Bromoquinazoline-2,4(1H,3H)-dione 97%
6-Bromoquinazoline-2,4(1H,3H)-dione

6-Bromoquinazoline-2,4(1H,3H)-dione, 97%


Purity: 97% CAS No.: 88145-89-5 Molecular Formula: C8H5BrN2O2 Formula Weight: 241.04 Physical Form: Solid Appearance: Off-white MDL No.: MFCD00462868

Formula: C8H5BrN2O2
MW: 241.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00462868
CAS Number: 88145-89-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR