Boc-4-(Fmoc-amino)-1-phenylalanine 95%
Supplier: Thermo Scientific Chemicals
H63543-03
H63543-06
AAH63543-03EA
89.11
CAD
AAH63543-03
AAH63543-06
Boc-4-(Fmoc-amino)-1-phenylalanine 95%
Boc-4-(Fmoc-amino)-1-phenylalanine
N-Boc-4-(Fmoc-amino)-L-phenylalanine, 95%
Purity: 95%
CAS No.: 114346-31-5
Molecular Formula: C29H30N2O6
Formula Weight: 502.56
Physical Form: Solid
MDL No.: MFCD00151889
Formula:
C₂₉H₃₀N₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00151889 CAS Number: 114346-31-5 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...