3-(2-Naphthyl)-D-alanine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-alpha-Amino-2-naphthalenepropionic Acid, H-D-2-Nal-OH
H63532-03
H63532-06
H63532-14
AAH63532-03EA
122.45
CAD
AAH63532-03
AAH63532-06
AAH63532-14
3-(2-Naphthyl)-D-alanine 95%
3-(2-Naphthyl)-D-alanine
3-(2-Naphthyl)-D-alanine, 95%
Purity: 95%
CAS No.: 76985-09-6
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 4352384
Optical Rotation: +12°(c= 1M HCl)
MDL No.: MFCD00038546
Formula:
C₁₃H₁₃NO₂ MW: 215.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038546 CAS Number: 76985-09-6 EINECS: 616-416-6 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...