H-Thr(tBu)-OMe·HCl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl O-(tert-butyl)-L-threoninate hydrochloride, O-tert-Butyl-L-threonine methyl ester hydrochloride, H-Thr(tBu)-OMe·HCl

H63529-03 H63529-06
AAH63529-03EA 62.41 CAD
AAH63529-03 AAH63529-06
H-Thr(tBu)-OMe·HCl 95%
H-Thr(tBu)-OMe·HCl

O-tert-Butyl-L-threonine methyl ester hydrochloride, 95%


Purity: 95% CAS No.: 71989-43-0 Molecular Formula: C9H19NO3.HCl Formula Weight: 225.71 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 4716005 MDL No.: MFCD00077111

Formula: C₉H₂₀ClNO₃
MW: 225.713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077111
CAS Number: 71989-43-0

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR