2,4-Dimethoxy-1-fluorobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H63528-03 H63528-06
AAH63528-03EA 142.52 CAD
AAH63528-03 AAH63528-06 AAH63528-MD
2,4-Dimethoxy-1-fluorobenzene 97%
2,4-Dimethoxy-1-fluorobenzene

1-Fluoro-2,4-dimethoxybenzene, 97%


Purity: 97% CAS No.: 17715-70-7 Molecular Formula: C8H9FO2 Formula Weight: 156.15 Flash Point: 105°(221°F) Physical Form: Liquid Density: 1.188 Appearance: Pale yellow MDL No.: MFCD00042265

Formula: C₈H₉FO₂
Density: 1.188
Flash Pt: 105 °C(221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042265
CAS Number: 17715-70-7
EINECS: 605-794-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR