3-(4-Thiazolyl)-L-alanine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63511-03
H63511-MD
AAH63511-03EA
504.02
CAD
AAH63511-03
AAH63511-MD
3-(4-Thiazolyl)-L-alanine 95%
3-(4-Thiazolyl)-L-alanine
3-(4-Thiazolyl)-L-alanine, 95%
Purity: 95%
CAS No.: 119433-80-6
Molecular Formula: C6H8N2O2S
Formula Weight: 172.20
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Pale yellow
MDL No.: MFCD00079677
Formula:
C₆H₈N₂O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD00079677 CAS Number: 119433-80-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...