(S)-(-)-2,3-Epoxy-1-propanol ≥99%, ee 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S)-(-)-Glycidol, (S)-Oxiran-2-ylmethanol
H63502-03
H63502-06
H63502-14
AAH63502-03EA
59.72
CAD
AAH63502-03
AAH63502-06
AAH63502-14
(S)-(-)-2,3-Epoxy-1-propanol ≥99%, ee 99%
(S)-(-)-2,3-Epoxy-1-propanol
(S)-(-)-Glycidol
Purity: 99+%, ee
CAS No.: 60456-23-7
Molecular Formula: C3H6O2
Formula Weight: 74.08
Storage Temperature: -30°C to -10°C
Flash Point: 66°(151°F)
Physical Form: Liquid
Density: 1.116
Refractive Index: 1.4330
Appearance: Pale yellow
Boiling Point: 66-67°/19mm
Beilstein Reference: 79783
Optical Rotation: -22°(c=1.35 in Chloroform)
UN No.: 2810
MDL No.: MFCD00074874
Formula:
C₃H₆O₂ Boiling Pt: 66…67 °C/19 mm Density: 1.116 Flash Pt: 66 °C(151 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00074874 CAS Number: 60456-23-7 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...