Methyl thiazole-2-carboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-thiazolecarboxylate
H63496-03
H63496-06
AAH63496-03EA
128.3
CAD
AAH63496-03
AAH63496-06
AAH63496-MD
Methyl thiazole-2-carboxylate 95%
Methyl thiazole-2-carboxylate
Methyl thiazole-2-carboxylate, 95%
Purity: 95%
CAS No.: 55842-56-3
Molecular Formula: C5H5NO2S
Formula Weight: 143.16
Physical Form: Solid
MDL No.: MFCD18075580
Formula:
C₅H₅NO₂S MW: 143.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18075580 CAS Number: 55842-56-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...