N-Formyl-L-methionine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: For-Met-OH, Formyl-L-methionine

H63493-03 H63493-06 H63493-MD
AAH63493-03EA 109.49 CAD
AAH63493-03 AAH63493-06 AAH63493-MD
N-Formyl-L-methionine 95%
N-Formyl-L-methionine

N-Formyl-L-methionine, 95%


Purity: 95% CAS No.: 4289-98-9 Molecular Formula: C6H11NO3S Formula Weight: 177.22 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00021033

Formula: C₆H₁₁NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00021033
CAS Number: 4289-98-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR