Fmoc-Oic-OH 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63491-03
H63491-MD
AAH63491-03EA
410.8
CAD
AAH63491-03
AAH63491-MD
Fmoc-Oic-OH 98%
Fmoc-Oic-OH
(2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid, 98%
Purity: 98%
CAS No.: 130309-37-4
Molecular Formula: C24H25NO4
Formula Weight: 391.46
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00191201
Formula:
C₂₄H₂₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00191201 CAS Number: 130309-37-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...