3-Nitro-D-phenylalanine 95%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Phe(3-NO₂)-OH, (R)-2-Amino-3-(3-nitrophenyl)propanoic acid

H63483-03 H63483-MD
AAH63483-03EA 341.28 CAD
AAH63483-03 AAH63483-MD
3-Nitro-D-phenylalanine 95%
3-Nitro-D-phenylalanine

3-Nitro-D-phenylalanine, 95%


Purity: 95% CAS No.: 169530-97-6 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Off-white to pale yellow MDL No.: MFCD04117829

Formula: C₉H₁₀N₂O₄
MW: 210.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117829
CAS Number: 169530-97-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR